Specifications
- Capacity:Up to 47.3 L (12.5 gal.)
- Cat. no.:56620-550
- Supplier No.:KIT411-01
Specifications
About this item
Overpack drum container is UN Rated for shipping waste after oil spill cleanup
Overpack is X-rated in Packing Groups I, II and III for shipping spill cleanup waste by land, sea or rail. Lightweight, 100% copolymer container resists chemicals and keeps contents clean and dry. Leakproof, twist-on lid is notched for easy removal or tightening with a 2x4 or pole. Large, color-coded label makes identification fast and accurate. Ledges molded into container make kit easy to move by forklift. Tamperproof seals help prevent pilfering of spill response supplies.
No wheels. UN1H2/X75/S, UN1H2/Y100/S.
Neatly prepacked container speeds access during a spill emergency. PIG Oil-Only Booms stop spreading spills; PIG Stat-Mat and Pillows absorb quickly. Absorbents meet NFPA 99 standards for static decay to allow use with fuels and flammables