Ultra-Precision NMR Sample Tubes, 100 Mhz, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Ultra-precise, accurate NMR sample tubes
Available in 17.8 cm (7") and 20.3 cm (8") lengths.
Customers Who Bought This Also Bought