Eurostar 20 Digital Electronic Overhead Stirrer, IKA® Works
# de catalogue 89412-482
Fournisseur: IKA Works
Spécifications
- Maximum Volume:15 L (3.96 gal.)
- Description:Eurostar 20 Digital Electronic Overhead Stirrer
- Permissible ambient temperature:5…40 °C
- Chuck range:0.5 - 10 mm
- Permissible relative humidity:80%
- Protection class DIN EN 60529:IP 40
- Electrical:115 V, 50/60 Hz
- Speed Range:0 - 2000 rpm
- Supplier No.:4442001
- Maximum Torque:20 Ncm
- Maximum Viscosity:10000 mPa·s
Spécifications
A propos de cet article
Laboratory stirrer designed for simple stirring tasks for quantities up to 15L (H₂O)
The Eurostar 20 Digital Electronic Overhead Stirrer features digital speed display, infinitely adjustable speed, push-through agitator shafts, overload protection, short-term overload operation, slim casing, quiet operation, and error code display.
The unit automatically adjusts the speed through microprocessor controlled technology within the speed range of 0/30–2000rpm. Safety circuits installed ensure automatic cut-off in anti-stall or overload conditions. Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed, even with changes in viscosities of the sample.
Certifications: CE marked.
Ordering information: Supplied with three-year manufacturer's warranty (2+1 with product registration). Unit includes chuck, chuck key, and power cord. Stirring elements, boss head clamps, stands, and other accessories are sold separately.
Customers Who Bought This Also Bought