SP Bel-Art Wet Surface Pen, Bel-Art Products, a part of SP
Catalogue # 47750-778
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Description:Wet surface pen
- Cat. no.:47750-778
- Supplier No.:133820000
Spécifications
À propos de cet article
Versatile "space pen" writes clearly on wet, greasy, and other hard to mark surfaces.
- Compatible for writing on otherwise hard-to-mark surfaces
- Can be used underwater or in extreme temperatures
- Writes on dry paper as a standard ballpoint pen
For use on wet labels, in water baths, liquid nitrogen, freezers at –20 °C (–4 °F), on filters while processing underwater-based buffers, for marking slots on Southern, Northern, and Western blots or on vertical surfaces.