Sawtooth Dispersing Tool for T25 Digital Homogenizer, IKA® Works
Fournisseur: IKA Works
Certificats
A propos de cet article
Dispersing tool for T25 Digital homogenizer
Sawtooth design creates a cutting and tearing action for shredding fibrous materials.
Made of high quality stainless steel.
Recommended for use with tissue and other fibrous materials.
Spécifications
- Catalog No:
- 89414-950
- 89414-948
- 89414-954
- 89414-952
- Model:
- S25N-10G-ST
- S25N-8G-ST
- S25N-25G-ST
- S25N-18G-ST
- Gap between rotor and stator:
- 0.35 mm
- 0.25 mm
- 0.5 mm
- 0.25 mm
- Stator Ø:
- 10 mm
- 8 mm
- 25 mm
- 18 mm
- Rotor Ø:
- 7.5 mm
- 6.1 mm
- 20 mm
- 13.4 mm
- Materials in contact with medium:
- PTFE, AISI 316L
- PTFE, AISI 316L
- PTFE, AISI 316L
- PTFE, AISI 316L
- Max. operating temperature:
- 180°C
- 180°C
- 180°C
- 180°C