ContactUsLinkComponent
PIG® Rip-and-Fit® HazMat Mat Roll, New Pig
PIG® Rip-and-Fit® HazMat Mat Roll, New Pig
Catalog # 89094-548
Supplier:  New Pig
Hazmat  Roll 15X16IN Diameter
PIG® Rip-and-Fit® HazMat Mat Roll, New Pig
Catalog # 89094-548
Supplier:  New Pig
Specifications
  • Description:
    Heavyweight
  • Dimensions:
    38.1 cm x 46 m (15" x 150')
  • Cat. no.:
    89094-548
  • Supplier No.:
    MAT343
Specifications
About this item
Absorbs the widest range of acids, bases and unknown liquids, even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide
Absorbs and retains most acids, bases and unknown liquids. Pink color is easily distinguishable to assure workers use the correct mat during a spill emergency. Perforated every 3-3/4" across width and every 10" across length; "rip" the perfect sized mat, then "fit" it into place.
Heavyweight construction is highly durable and absorbent for demanding tasks and large-volume cleanups.
Chemical-resistant mat won't degrade or cause a dangerous reaction upon contact with corrosive spills. Extra perforations in our Rip-and-Fit Mat allow you to easily "rip" this PIG Mat and "fit" it to your application without cutting. No matter how you rip it, this mat features the proven eight-layer, 100% polypropylene construction of PIG Mat. Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly-absorbent, fine-fiber construction won't leave behind liquids or fiber residue
Customers Who Bought This Also Bought