PIG® Latching Drum Lid for Open-Head Steel Drums, New Pig
Fournisseur: New Pig
Certificats
A propos de cet article
Latching handle easily opens and closes with one hand.
- Nitrile gasket and locking mechanism seal lid, reduces vapor emissions keeps drum contents dry and secure
- Easy-access lid closes securely to help you meet haz-waste and closed-container regs
Hinge stops keep funnel lid upright when open. Durable, chemical-resistant powder coat helps ensure long service life. Lockable latch helps prevent unauthorized access. Hinge pin is made for easy lid removal without loosening band around drum. Hinged with one hand latch.
BodyTextAccessories: Replaces gasket on model DRM659 (PIG Latching Drum Lid). Includes gasket and adhesive.
Customers Who Bought This Also Bought