Open-Head UN Rated Poly Drum, New Pig
Fournisseur: 2 D 2 S.A. DE C.V. MX
Certificats
A propos de cet article
Rigid, high-density polyethylene is lightweight and won't rust
- Rigid, high-density polyethylene is lightweight and won't rust
- Compatible with a wide range of materials, liquids and chemicals
- Made of USDA- and FDA-Certified and Approved resins for direct containment of food products
Lever lock band can be wire-sealed to prevent theft. Ideal for storage of chemicals, food products and pharmaceuticals. UN Rated for shipping solids.
Compatible with a wide range of materials, liquids and chemicals. Made of USDA and FDA Certified and Approved resins for direct containment of food products.
Customers Who Bought This Also Bought