PIG® Heavy Fluids Absorbent Mat Pad, New Pig
# de catalogue 56620-546
Fournisseur: New Pig
Spécifications
- Description:Heavyweight, 50 Pads
- Cat. no.:56620-546
- Dimensions:45.7 cm x 55.9 cm (18" x 22")
- Supplier No.:MAT209
Spécifications
A propos de cet article
Thirsty air pockets absorb even the stickiest heavy liquids with only slight pressure
Excellent for cleaning up paint spills, glues, resins, liquid chocolate, syrup and varnish. Quickly absorbs thick, high-viscosity liquids that conventional absorbents can't. Can also be used to filter metal chips and other large debris from liquids. Pads are excellent for cleaning up smaller spills of thick liquids.
Customers Who Bought This Also Bought