Pop-Up Drum Fill Gauge, New Pig
Fournisseur: New Pig
Certificats
A propos de cet article
Durable gauge helps prevent overfilling by popping up when drum is near capacity.
- Corrosion-resistant polyethylene withstands most liquids
- Highly-visible red and yellow offers easy indication
Avoid overflows and downtime by quickly checking a drum's liquid level-no guessing or messy dipsticks. For 55 gal. vertical drums with 0.75" NPT bung.
Customers Who Bought This Also Bought