PIG® Steel Loading Ramp, New Pig
# de catalogue 56620-616
Fournisseur: New Pig
Spécifications
- Description:For 4" to 51/4" H steel or poly spill containment decks
- Cat. no.:56620-616
- Supplier No.:PAK538
Spécifications
A propos de cet article
Heavy-duty steel ramps are built strong for big loads. Skip the forklift and roll your drums up a ramp.
- Heavy-duty steel construction is strong and durable
- Adjustable legs for use with a range of pallet and deck heights
- Edge guards guide drum trucks and help prevent them from rolling off the sides
- Lip of ramp secures to edge of pallet or deck
Note: Ramp must be detached to close doors. Diamond plate surface provides traction for added safety.
Customers Who Bought This Also Bought