PIG® Single Drum Container, New Pig
# de catalogue 56620-126
Fournisseur: New Pig
Spécifications
- Description:Holds One 55-gal. Steel or Poly Drum
- Cat. no.:56620-126
- Load Capacity:544.3 kg (1200 lbs.)
- Sump Capacity:265 L (70 gal.)
- Supplier No.:DRM373
Spécifications
A propos de cet article
Low-density polyethylene construction resists UV rays, rust, corrosion and most chemicals
- Sump is large enough to contain a leaking or fully ruptured 55-gallon drum
- Top collar lets you easily move container with a forklift and holds up to a 1,200-lb. load
Translucent white sidewalls allow easy visual inspection.
Customers Who Bought This Also Bought