PIG® Heavy-Duty Poly Modular Spill Deck, New Pig
# de catalogue 56620-156
Fournisseur: New Pig
Spécifications
- Description:Holds Four 55-gal. Steel or Poly Drums
- Cat. no.:56620-156
- Load Capacity:50000 lbs.
- Sump Capacity:35 gal.
- Supplier No.:PAK466
Spécifications
A propos de cet article
Heavy-duty deck withstands extra-heavy loads; Excellent for storing or servicing heavy equipment that is prone to leaks
- Low 4.25" height is easy to load or unload; pumps and funnels are easy to access
- Catches leaks, drips and spills to help keep your storage areas clean and safe
- Low-density polyethylene (LDPE) construction resists UV rays, rust, corrosion and most chemicals
- Textured grating adds traction and lifts out for easy sump access
Unique "flow-through" sump design lets liquid pass freely from one sump to another to help you comply with containment regulations. Modular design features exclusive bulkhead fittings and predrilled ports to connect to other decks; ready to use as soon as they arrive. Mix and match the sections you need to fit your space and help meet sump capacity requirements. Special tools and hardware to connect sections is included with each system.
Customers Who Bought This Also Bought