Violet Red Bile Agar, CRITERION™, Hardy Diagnostics
Fournisseur: HARDY DIAGNOSTICS CA
Certificats
A propos de cet article
CRITERION™ Violet Red Bile Agar is recommended for the detection of coliforms in food or dairy products.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Certifications: BSE Free
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.