Lactobacilli MRS Agar, Hardy Diagnostics
Fournisseur: HARDY DIAGNOSTICS CA
Certificats
A propos de cet article
Lactobacilli MRS Agar is recommended for the isolation, enumeration and cultivation of Lactobacillus spp.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.