HardyVal CSP Complexity Kit
Fournisseur: HARDY DIAGNOSTICS CA
Certificats
A propos de cet article
A self-contained ready-to-use kit with all the products needed to easily assess the risk of microbial contamination of your CSPs (Compounded Sterile Preparations) according to the new USP 27, Chapter 797 requirements for media-fill challenge testing
Kit performs one complete test for one technician. Includes test results log sheet.
Customers Who Bought This Also Bought