Bile Salts, #3, CRITERION™, Hardy Diagnostics
Fournisseur: HARDY DIAGNOSTICS CA
Certificats
A propos de cet article
CRITERION™ Bile Salts #3 is specifically formulated for use in MacConkey Agar, SS Agar, and Violet Red Bile Agar
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.