Cutters, Tubing, Excelta
Fournisseur: Excelta
Certificats
A propos de cet article
Gives a nice clean cut to a variety of soft tubing materials.
- Medical 'clean-cut' tubing cutter
- Replaceable blades
- Ergonomic grips and dual leaf springs
The 2922 Series and the 51-T medical tubing cutter give a nice clean cut to a variety of soft tubing materials. The blades on the 2922 series are replaceable and the grips are soft cushioned foam and have a surface resistivity of 10¹⁰ ohms/sq.
Customers Who Bought This Also Bought