Intellitweeze R-C-L Meter, Excelta
# de catalogue 89411-972
Fournisseur: Excelta
Spécifications
- Description:Five Star Quality
- Cat. no.:89411-972
- Supplier No.:TM-500
Spécifications
A propos de cet article
The IntelliTweeze is a quick and easy way of testing and troubleshooting many types of components
With the use of only one hand, this lightweight unit will quickly measure resistance, capacitance, inductance, continuity and polarity.
Customers Who Bought This Also Bought