Solid State Voltage Controller, 0 – 120 V at 10 Amps, Ace Glass Incorporated
# de catalogue 89076-332
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Length:17.8 cm (7")
- Description:Solid State Voltage Controller, 120 V, 50/60 Hz, 1 PH (Input), 0 – 40 V or 0 – 120 V (Selectable Output Range)
- Depth:8.9 cm (31/2")
- Height:10.2 cm (4")
- Cat. no.:89076-332
- Supplier No.:12087-10
Spécifications
A propos de cet article
This Solid State Voltage Controller offers variable control from zero output to 95% line voltage.
A voltage-limit, rear-mounted center-off switch is used to select a 40 V or 120 V maximum output. With 0 – 10 ammeter, and 5% accuracy. Warning glow light mounted next to correcting switch which lights when dangerous reversed wiring condition exists. Simply flip switch to extinguish light and correct condition. Easy-access 10 amp fuse type 3 AG (rear mounted). Line voltage pilot light. Standard 1.8 meter heavy duty neoprene three-wire power cord (grounded to case) with NEMA plug. Input 120 volts, 60 cycles.
Weight: 794 g (1.75 lbs.)
Caution: As this is a solid state transformerless line voltage controller, it is NOT recommended for heaters rated less than 120 V.
Customers Who Bought This Also Bought