Heat Gun Mite®, Ace Glass Incorporated
# de catalogue 89076-310
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Heat Gun Mite®
- Electrical:5.4 A
- Voltage:120 V
- Supplier No.:12072-20
Spécifications
A propos de cet article
Economical, compact, flameless heat gun designed for heat shrinkage as well as countless other industrial applications
Input is 120V, 5.4 amps. Supplied with 2.1m, three-conductor grounded cord. Unit consists of heat gun with silver nozzle and deflector adapter.
Customers Who Bought This Also Bought