Heat Gun/Dryer, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Heavy-duty, flameless heat gun
Compact gun-type housing is constructed from die-cast aluminum and features a 30mm diameter, screen-capped nozzle. Adjustable air-intake shutter gives exact degree of heat desired. Three-way switch provides the option of hot, cold, or off at the flick of a finger. Supplied with 2.4m, three-wire molded plug neoprene cord and adjustable “slide-on” base.
Customers Who Bought This Also Bought