Varitemp® Heat Gun, Ace Glass Incorporated
# de catalogue 89076-308
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Varitemp® Heat Gun
- Cat. no.:89076-308
- Supplier No.:12070-10
Spécifications
A propos de cet article
Heavy-duty, flameless heat gun features an exclusive locking electronic temperature control that enables the user to dial in the precise temperature for the job
The control locks in place to allow the same setting to be used repeatedly. Approximate temperature range at nozzle is ambient to 400°C (752°F). The heat gun features a 13/16" nozzle with a powerful turbo fan that is controlled by a three-way switch with guard. Reinforced ceramic heating element with double-jacketed heater housing for protection. Rugged, die-cast aluminum housing with externally replaceable carbon brushes. Supplied with a 243.8cm (8') neoprene cord with three-wire ground plug.
Customers Who Bought This Also Bought