Mantles, Fabric/Cloth, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Heating mantles are designed for use with reaction flasks
Upper temperature limit is 450°C (842°F). A 122cm (4') cord and locking connector are included. Units must be operated through a temperature controller.
Certifications: CSA approved.
Customers Who Bought This Also Bought