Gas Collecting Tube, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Standard collecting tube with glass stopcocks on each end and capillary tubing for connecting to rubber tubing and a side syringe sampling port with septum.
- Fabricated from borosilicate glass
Gas collecting tube 89058-814 is a standard collecting tube supplied with 1:5 taper PTFE stopcock plugs in place of glass plugs on each end and a side syringe sampling port with septum.
For replacement septum, see item 89064-962.
Customers Who Bought This Also Bought