Gas Manifold, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Designed for use as a portable gas analysis apparatus
Manifolds are available with either three or four straight bore 2 mm glass stopcocks and one three-way stopcock with back port, fabricated on capillary tubing. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought