High Temperature Dry Block Heater, BT5D, Grant Instruments
Fournisseur: GRANT USA INC.
Certificats
A propos de cet article
High temperature digital block heaters with single fixed aluminium block
- Operating temperature range is ambient +10 to 400°C
- Temperature stability at 300°C is ±0,5°C and temperature uniformity is ±1% (maximum temperature difference between tubes: 2°C at 200°C, 3°C at 300°C, 4°C at 400°C)
- Timer for operation from 1 to 9999 min., or continuous operation
- Ideal for use at high temperatures - avoids the use of oil and laboratory remains steam-free
- Adjustable overtemperature cut-out
- Choice of two models with different block capacities
The BT5D is ideal for any application requiring heating in a dry block up to 400ºC.
Units feature microprocessor control, digital temperature settings and display, high and low alarms, and a controlled heating rate. Heaters have LED with 0,1°C setting resolution and 1°C display resolution.
Certifications: CE Approved.
Ordering information: Choice of two models: BT5D-16 for 38 x Ø 16 mm tubes and BT5D-26 for 22 x Ø 26 mm tubes. Other sizes also available; please contact your Avantor representative for more information.
Spécifications
- Catalog No:
- 89498-130
- 71003-686
- Timer:
- 1 to 9999 mins
- 1 to 9999 mins|1 to 9999 Mins.
- Uniformity:
- ±0.01°C
- ±0.01°C|0.01°C
- Alarm:
- High and Low
- High and Low
- Electrical:
- 120 V
- 110-230V
- Stability:
- ±0.5 (up to 300°C)
- ± 0.5 (up to 300°C), stability to DIN 58966|±0.5 (up to 300°C)