Sample Flask, with Standard Taper Joint, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Flasks feature a standard taper joint on the side arm in the vertical position
The 5mL size is tube style, and the 25mL size is bulb style. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought