A propos de cet article
TrippNT® extra-wide compact cart is a 27" wide food grade high density polyethylene lab cart in six colors with built-in tray slides under counters, benchtops, and fume hoods.
- Non-metal rust proof polymer construction performs well in harsh, humid, corrosive environments
- Six colors
- Four full-extension drawers
- One key locks all drawers
- Built-in tray top and handle for push pull control
- 150 lb. weight capacity
- Decontaminates with bleach or aqueous solutions
- Fully assembled
- American made by TrippNT
- Total replacement lifetime guarantee
The Extra Wide Compact Drawer Cart creates storage for; glassware, tools, utensils, reagents, chemicals, pens, pippettes, tweezers, log books, documents, bottles and other laboratory supplies.
Features a built-in tray top. Four full-extension drawers (pulls all the way out to fully access drawer contents). Clearance from bottom of drawer to drawer above is 4½". Industrial drawer slides keep drawer closed while cart is moving. Stong, ergonomic, high density polyethylene handles.
Perfect for labs, clinics, hospitals, anesthesia supplies, doctor's offices and similar industries to add extra work space, storage, and mobility. Fits comfortably under tables, counters, benches, fume hoods, and biosafety cabinets.
Spécifications
- Catalog No:
- 10805-268
- Material:
- Food Grade High Density Polyethylene, Natural White; ABS (Acrylonitrile butadiene styrene) drawers; PVC drawer fronts
- Storage capacity:
- 4.7 cu. ft.
- Drawer dimensions:
- 24½W×4½H×15½" D
- Tray dimensions:
- 25¼×3¼×15⅝", top
- Width:
- 25⅜"
- Depth:
- 18⅛"
- Height:
- 27⅝"
- Weight:
- 67 lbs.