A propos de cet article
Heavy duty food grade high density polyethylene lab medical cart in many different colors with built-in tray top.
- Available in 13 colors
- Tall space saving lab cart with 6 drawers only 16 inches wide
- A new innovative option to traditional metal carts
- Non-metal rust proof polymer construction performs well in harsh, humid, corrosive environments
- 4 inch Total Lock Casters (locks rotation and swivel)
- Decontaminates with bleach or aqueous solutions
- Extra wide base for stability
- 6 full-extension drawers (4 small, 2 large)
- 1 key locks all drawers
- 1 piece body construction for strength
- 3 inch built-in Tray Top for storage
- Durable chemical resistant
- 200 pound weight capacity
- Fully assembled
- American Made by TrippNT
- Total replacement Lifetime Guarantee
Storage for small or bulky items and heavy-duty, one piece construction adds strength and security for trauma, ER, surgical, anesthesia, critical care, and any hospital, lab or care center use. Unique tall slender design creates maximum storage in a small space.
Heavy duty, substantial, meant-to-be-used mobile cabinet features 6 full-extension drawers (pulls all the way out to fully access drawer contents). 4 small drawers that are 11-1/2 inches wide by 4-1/2 inches high and 15-1/2 inches deep. 2 large drawers that are 11-1/2 inches wide by 9 inches high and 15-1/2 inches deep.
Industrial drawer slides keep drawer closed while cart is moving. Strong, ergonomic, high density polyethylene handles.
The Locking Tall 6 Drawer Polyethylene Polymer Non-Metal Storage Cart creates storage for syringes, needles, alcohol wipes, swabs, glassware, tools, utensils, reagents, chemicals, gloves, safety glasses, pipettes, tweezers, log books, documents, bottles and other emergency supplies. 1 key lock feature adds extra security for storing medication and other supplies that need to be kept safe. Features a 12 3/4 by 3 1/4 by 15 5/8 inch built-in Tray Top.
Perfect for labs, clinics, operating rooms, ER's, patient supplies, hospitals, doctor's offices and similar industries to add extra space, storage, and mobility. The TrippNT Locking Tall 6 Drawer Polyethylene Cart adds security and maximum storage for any hospital or care facility.
Spécifications
- Catalog No:
- 89408-468
- 10754-646
- Storage capacity:
- 4.1 cu. ft.
- 4.1 cu. ft.
- Caster type:
- 4" Diameter total locking polypropylene casters (locks rotation and swivel)
- 4" Diameter total locking polypropylene casters (locks rotation and swivel)
- Drawer dimensions:
- Small Drawer: 111/2" x 41/2" x 151/2"; Large Drawer: 111/2" x 9 x 151/2", internal
- Small Drawer: 111/2" x 41/2" x 151/2"; Large Drawer: 111/2" x 9 x 151/2", internal
- Tray dimensions:
- 123/4" x 31/4" x 155/8", Top
- 123/4" x 31/4" x 155/8", Top