Flask Heater, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Specially designed glass/fiber cloth mantle with bottom opening and zipper for use with flasks to allow valve to protrude below heater
Upper temperature limit is 450 °C (842 °F). Must be operated through a temperature controller.
Certifications: CSA approved.
Ordering information: Flask heater is supplied with 122 cm (4') cord and locking connector.
Customers Who Bought This Also Bought