A propos de cet article
The Barnstead GenPure Pro water purification systems provide up to 200L/day of Type 1, ultrapure (18.2 megohm) water from pre-treated feed water
The GenPure Pro systems offer highly versatile mounting options. The units can be mounted on the wall, on the bench or under the bench. In addition the GenPure Pro offer highly flexible dispensing with a its flexible dispensing arm to accommodate the filling of a wide range of vessels.
The GenPure Pro systems are offered in a variety of configurations to meet a wide range of applications needs. The GenPure Pro systems are ideal for atomic absorption, ion chromatography or other analytical procedures. The GenPure Pro UV systems are ideal for inorganic and organic trace analysis, HPLC, or ICP-MS.
The GenPure Pro UF systems are ideal for microbiology, IVF, or monoclonal antibody applications. The GenPure Pro UV/UF systems are ideal for molecular biology, PCR, DNA work, cell or tissue culture media. In addition the UV and UV/UF models can be ordered with an optional TOC monitor for applications that require TOC monitoring in addition to TOC removal. Both models with TOC monitoring include UV intensity monitoring to ensure TOC readings are accurate.
All GenPure Pro systems include feed water monitoring to protect your consumables, built in leak detection, quick disconnects to make changing consumables easy, all core consumables and tubing, and RS232 port.
Spécifications
- Catalog No:
- CA89414-748
- 89414-744
- W×D×H:
- 372×330×615 mm (14.6×13×24")
- 372×330×615 mm (14.6×13×24")
Spécifications
Informations connexes
In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWR[email protected] for further details.