CritiCore Indura Unisex Flame Resistant Lab Coats
Fournisseur: VWR International
Certificats
A propos de cet article
CritiCore has developed flame-resistant lab coats to provide the protection required in many of today's high-tech laboratories and research facilities.
- Flame-resistant identification patches sewn on the back and both sleeves for easy recognition
- Quick-release snap front for fast removal
- Safety-sleeve cuff with tunneled elastic
- Two oversized waist pockets and one breast pocket with a convenient pencil slot
- Vent openings on the sides allow access to pant pockets
These coats are made with 100% cotton fabric, durable enough to withstand industrial laundering while maintaining fire-resistant properties for the life of the garment.
Certifications: CAT I
Ordering information: Available only in blue. Coats in sizes X-Small to XX-Large are stocked. For sizes 3X-Large to 6X-Large, extended lead time may be required. Contact a Avantor sales representative for more information.
Spécifications
- Catalog No:
- 89426-882
- 89426-884
- 89426-890
- 89426-892
- 89426-886
- 89426-888
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Gender:
- For men
- For men
- For men
- For men
- For men
- For men
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton