A propos de cet article
Enhance and simplify your low-temperature laboratory work with these low temperature coolers. Available as immersion probe and flow-through models, these units provide rapid, low-cost cooling of liquids to temperatures as low as -100 °C. Typical applications include cooling exothermic reactions, freeze point determinations, freeze drying, impact testing, lyophilization and vapor and solvent trapping.
Flow-through Style Coolers offer an extremely economical alternative to tap water cooling. Use with heated circulating baths to reach temperatures below ambient. Use with refrigerated circulating baths to increase cooling capacity.
Flow-through style coolers offer an extremely economical alternative to tap water cooling. Use with heated circulating baths to reach temperatures below ambient. Use with refrigerated circulating baths to increase cooling capacity.
Immersion probe style coolers are available with different fixed temperatures. Excellent for trapping and Dewar-type applications, these coolers are designed to run at maximum cooling and reduce the expense of using dry ice or liquid nitrogen. A flexible hose allows convenient placement of the cooling probe.
Spécifications
- Catalog No:
- 89400-962
- 89400-958
- 89400-966
- 89400-968
- 89400-964
- Electrical:
- 240V, 50Hz
- 240V, 50Hz
- 240V, 50Hz
- 240V, 50Hz
- 240V, 50Hz