Amplitude™ Knitted Wipes, Heavy Weight
Fournisseur: Contec
Certificats
A propos de cet article
The Amplitude heavy weight wipe is constructed from 100% textured polyester knit.
Extremely low in both particles and fibers. Good sorbency with solvents. Abrasion and chemical resistant. Ideal for general purpose cleaning.
- Extremely low in both particles and fibers
- Good sorbency with solvents
- Abrasion and chemical resistant
- Ideal for general purpose cleaning
The Amplitude heavy weight wipe is constructed from 100% textured polyester knit.
The wipe is laser cut, which bonds the fibers at the edges of the wipe. This creates a very clean, nonabrasive edge which allows 100% utilization of the wipe surface.
Spécifications
- Catalog No:
- 89128-800
- 94002-682
- Material:
- Polyester knit
- Polyester knit