ContactUsLinkComponent
ORGANELLE-ID-RGB® Reagent II, Enzo Life Sciences
ORGANELLE-ID-RGB® Reagent II, Enzo Life Sciences
# de catalogue 89166-072
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Organelle-Id* Rgb Reagent Ii 200 ul
ORGANELLE-ID-RGB® Reagent II, Enzo Life Sciences
# de catalogue 89166-072
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Reagent II ORGANELLE-ID-RGB®
  • Size:
    200 µL
  • Purity:
    ≥93% (HPLC)
  • Cat. no.:
    89166-072
  • Supplier No.:
    53008-C200
Spécifications
A propos de cet article
Mixture of fluorescent dyes for detection of lysosomes, endoplasmic reticulum and nucleic acids.
The Organelle-ID™ RGB reagent II is a mixture of cell-permeable red fluorescent lysosomal dye, green fluorescent endoplasmic reticulum dye and blue fluorescent nucleic acid dye. The staining pattern arising from the combination of these three dyes permits visualization of the target organelles by fluorescence/confocal microscopy. The reagent, supplied as a 500X solution, is sufficient for 1000 microscopy assays. The single-tube format makes this multi-organelle stain reagent easy to use.
Ordering information: Quantity: 200 μl (for 1000 microscopy assays).
Customers Who Bought This Also Bought