ContactUsLinkComponent
NUCLEAR-ID® Red/Green Cell Viability Reagent, Enzo Life Sciences
NUCLEAR-ID® Red/Green Cell Viability Reagent, Enzo Life Sciences
# de catalogue 89166-068
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Nuclear-Id* Red/Green Cell Viability Reagent 100 ul
NUCLEAR-ID® Red/Green Cell Viability Reagent, Enzo Life Sciences
# de catalogue 89166-068
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Red/Green Cell Viability Reagent NUCLEAR-ID®
  • Size:
    100 µL
  • Purity:
    ≥93% (HPLC)
  • Cat. no.:
    89166-068
  • Supplier No.:
    53006-C100
Spécifications
A propos de cet article
Cell viability dye.
The Nuclear-ID™ Red/Green cell viability reagent (Prod. No. ENZ-53006) is a mixture of a red fluorescent cell-permeable nucleic acid dye and a green fluorescent cell-impermeable nucleic acid dye that is suited for staining dead nuclei. The staining pattern arising from the simultaneous combination of these two dyes permits determination of live and dead cell populations by fluorescence/confocal microscopy. The reagent, supplied as a 1000x solution, is sufficient for 1000 microscopy assays. The single-tube format makes this cell viability reagent easy to use. It leaves the cytoplasm unstained, potentially allowing visualization of other cell markers.
Ordering information: Quantity: 100 μl (for 1000 microscopy assays).
Customers Who Bought This Also Bought