ContactUsLinkComponent
ORGANELLE-ID-RGB® III Assay Kit, Enzo Life Sciences
ORGANELLE-ID-RGB® III Assay Kit, Enzo Life Sciences
# de catalogue 89165-928
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Organelle-Id Rgb* Iii Assay Kit 1 Kit
ORGANELLE-ID-RGB® III Assay Kit, Enzo Life Sciences
# de catalogue 89165-928
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Assay Kit
  • Size:
    100 Assay
  • Storage conditions:
    –20 °C (Short Term), –80 °C (Long Term)
  • Application:
    Fluorescence Microscopy, Fluorescent Detection
  • Cat. no.:
    89165-928
  • Supplier No.:
    51032-K100
Spécifications
A propos de cet article
The Organelle-ID RGB™ III assay kit contains Golgi apparatus-selective, endoplasmic reticulum-selective and nucleus-selective dyes suitable for live cell staining.
  • Highly resistant to photo-bleaching, concentration quenching and photoconversion
  • Identify modulators of secretory membrane traffic and organelle integrity
The green dye component of the Organelle-ID RGB™ III detection reagent is more faithfully localized to the Golgi apparatus, with minimal staining of the endoplasmic reticulum
Organelle-ID RGB™ III assay kit is validated with human cervical carcinoma cell line HeLa, human T-lymphocyte cell line, Jurkat, canine kidney cell line MDCK, and human bone osteosarcoma epithelial cell line U2OS. The kit should also be suitable for identifying Golgi body and endoplasmic reticulum perturbing agents and thus can be a useful tool for examining the transport and recycling of molecules from the GA to ER in cellular secretory pathways.
The red dye component of the reagent stains the endoplasmic reticulum with high fidelity and is specifically designed for use with green fluorescing probes.
Delivery: Kit includes ORGANELLE-ID-RGB® Reagent III (lyophilized) (1 vial), 10X Assay Buffer 1 (15 mL), 50X Assay Buffer 2 (1.2 mL).
Customers Who Bought This Also Bought