ContactUsLinkComponent
MITO-ID® Membrane Potential Cytotoxicity Kit, Enzo Life Sciences
MITO-ID® Membrane Potential Cytotoxicity Kit, Enzo Life Sciences
# de catalogue 89165-900
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Mito-Id* Membrane Potential Cytotoxicity Kit For Microplates 1 Kit
MITO-ID® Membrane Potential Cytotoxicity Kit, Enzo Life Sciences
# de catalogue 89165-900
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    1 Kit
  • Cat. no.:
    89165-900
  • Supplier No.:
    51019-KP002
Spécifications
A propos de cet article
A Real-time Mitochondrial membrane Potential Assay with Superior Sensitivity
Mito-ID® Membrane Potential Cytotoxicity Kit enables monitoring of mitochondrial potential changes using a simple fluorescence microplate reader.
The Mito-ID® Membrane Potential Cytotoxicity Kit measures fluctuations in mitochondrial membrane potential (MMP) utilizing a cationic dual-emission dye that exists as green fluorescent monomers in the cytosol, and accumulates as orange fluorescent J-aggregates in the mitochondria. Mitochondria having a low membrane potential will accumulate low concentrations of dye and will exhibit green fluorescence while more highly polarized mitochondria will exhibit orange-red fluorescence. Cells exhibit a shift from orange to green fluorescence as mitochondrial function becomes increasingly compromised. The kit is a unique HTS assay that monitors mitochondrial membrane potential in real-time without the need for washes or medium removal.
Mechanism of Action
The basic chemical structure of the dye consists of highly conjugated moieties that extensively delocalize a positive charge thus allowing electrophoretic uptake toward the negatively charged matrix phase of the polarized inner mitochondrial membrane. The dye is capable of entering selectively into mitochondria wherein it changes its color reversibly from green to orange as membrane potential increases (dual-emission potential probe). This photophysical property is due to the reversible formation of J-aggregates upon membrane polarization that causes shifts in emitted light from ~530nm (the emission of the monomeric dye) to 590 nm (the emission of the J-aggregate form) when excited at 490 nm. As a consequence, mitochondria having a low membrane potential will accumulate low concentrations of dye and will exhibit green fluorescence while more highly polarized mitochondria will exhibit orange fluorescence.
Ordering information: Quantity: For 2 x 96-well microplates Contents: Mito-ID® MP Detection Reagent, 200 μL
CCCP Control, 100 μL
10X Assay Buffer 1: 2.5 mL
50X Assay Buffer 2: 0.5 mL
Customers Who Bought This Also Bought