ContactUsLinkComponent
NUCLEAR-ID® Red Cell Cycle Kit (GFP-CERTIFIED®), Enzo Life Sciences
NUCLEAR-ID® Red Cell Cycle Kit (GFP-CERTIFIED®), Enzo Life Sciences
# de catalogue 89165-878
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
NUCLEAR-ID® Red Cell Cycle Kit (GFP-CERTIFIED®), Enzo Life Sciences
# de catalogue 89165-878
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    1 Kit
  • Cat. no.:
    89165-878
  • Supplier No.:
    51008-100
Spécifications
A propos de cet article
Suitable for DNA studies in live, permeabilized and fixed cells for normal cell lines and cell lines exhibiting multiple ploidy levels.
Enzo Life Sciences GFP-Certified™ Nuclear-ID™ Red Cell Cycle Analysis Kit provides a convenient approach for studying the induction and inhibition of cell cycle progression by flow cytometry. The kit is suitable for (1) determining the percentage of cells in a given sample that are in G0/G1, S and G2/M phases, as well as to quantify cells in the sub-G1 phase, and (2) DNA studies in live, permeabilized and fixed cells for normal cell lines and cell lines exhibiting multiple ploidy levels. A control cell cycle perturbation agent, Nocodazole, is provided for monitoring changes in cell cycle dynamics. Potential applications for live-cell studies are in the determination of cellular DNA content and cell cycle distribution for the detection of variations in growth patterns, for monitoring apoptosis, and for evaluating tumor cell behavior and suppressor gene mechanisms.
Ordering information: Quantity: 100 assays Contents: Nuclear-ID™ Red Cell Cycle Detection Reagent, 200 µL
Nocodazole Control, 10 µL
10X Assay Buffer, 15 mL
Customers Who Bought This Also Bought