ContactUsLinkComponent
Bio-16-ddUTP . tetralithium salt, Enzo Life Sciences
Bio-16-ddUTP . tetralithium salt, Enzo Life Sciences
# de catalogue 89165-830
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Bio-16-Ddutp . Tetralithium Salt 25 Nmol
Bio-16-ddUTP . tetralithium salt, Enzo Life Sciences
# de catalogue 89165-830
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Bio-16-ddUTP Tetralithium Salt
  • Size:
    25 nmoL
  • Purity detail:
    Contains \<5% Bio-16-ddUDP.
  • Purity:
    ≥93% (HPLC)
  • Concentration:
    1mM
  • Formulation:
    Liquid. Solution in water.
  • Supplier No.:
    42813
Spécifications
A propos de cet article
Bio-16-ddUTP (Biotin-16-2’,3’-dideoxyuridine-5’-triphosphate) can be used for 3’-end labeling of single-stranded or double-stranded DNA. It is especially useful as a substrate for terminal transferase for the 3’-terminal addition of a single Bio-16-ddUMP. Biotin-labeled oligonucleotides produced by this reaction are suitable as hybridization probes.
Biotin-labeled cDNA produced by this reaction can be used in nucleic acid array assays. Biotin-labeled DNA in hybridization assays can be detected by a reporter molecule linked to streptavidin, avidin, or and anti-biotin antibody. Such a complex can be detected directly, e.g. by the excitation of a fluorophore conjugated to streptavidin, or indirectly, e.g. using an enzyme conjugate that can produce an insoluble colored precipitate. Bio-16-ddUTP is also a substrate for Taq DNA Polymerase and other DNA polymerases.
Customers Who Bought This Also Bought