ContactUsLinkComponent
BIO-PROBE® Adenovirus Probe, Enzo Life Sciences
BIO-PROBE® Adenovirus Probe, Enzo Life Sciences
# de catalogue 89165-722
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
BIO-PROBE® Adenovirus Probe, Enzo Life Sciences
# de catalogue 89165-722
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    BIO-PROBE® Adenovirus Probe
  • Size:
    2 µg
  • Concentration:
    20µg/ml
  • Formulation:
    Liquid. In 10mM TRIS HCl, pH 7.5, containing 1mM EDTA.
  • Cat. no.:
    89165-722
  • Supplier No.:
    40834
Spécifications
A propos de cet article
The Adenovirus BioProbe® labeled probe is prepared by nick translation of cloned fragments of the adenovirus 5 genome.
The fragments constitute a total of 20 kb of DNA, approximately 56% of the genome. The probe is specific for adenovirus and, in addition to hybridization to adenovirus 5 sequences, the probe will cross hybridize with adenovirus types 4, 7, 11, 20, 40 and 41 due to their considerable sequence homology. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).
Customers Who Bought This Also Bought