ContactUsLinkComponent
Ubiquitin Conjugating Kit (HeLa Lysate-Based), Enzo Life Sciences
Ubiquitin Conjugating Kit (HeLa Lysate-Based), Enzo Life Sciences
# de catalogue 89165-450
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Ubiquitin Conjugating Kit (HeLa Lysate-Based), Enzo Life Sciences
# de catalogue 89165-450
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    1 Kit
  • Cat. no.:
    89165-450
  • Supplier No.:
    UW9915-0001
Spécifications
A propos de cet article
1. Ubiquitinylation of specific endogenous HeLa lysate proteins, followed by their immediate detection/analysis using antibodies to the protein/s of interest, indicating a particular protein is a substrate for the ubiquitin-proteasome pathway. 2. Ubiquitinylation of proteins of interest contained in exogenously added expression culture/cell extracts or tissue lysates/extracts, followed by their immediate detection/analysis or isolation/purification for use in subsequent experiments. 3. Modification of proteins using ubiquitin derivatives or mutants for improved detection/analysis or investigation of alternative (non-proteasomal) ubiquitin related pathways in subsequent experiments. For example, biotinylated-ubiquitin (Prod. No. BML-UW8705), for sensitive detection
For the generation of ubiquitin conjugated proteins. HeLa S100 lysate facilitates controlled ubiquitin conjugation of substrate proteins (exogenous or endogenous) of interest through the ubiquitin cascade. Conjugate formation can be detected and monitored by Western blotting using the highly sensitive ubiquitin-conjugate specific antibody supplied and/or antibodies for specific target proteins. Modified proteins can be subjected to further purification prior to their use in subsequent experiments if required. Suggested uses: Generation of ubiquitin conjugated proteins. Exogenous or endogenous HeLa lysate proteins (tagged/radio-labeled/immuno-detectable) can be ubiquitinylated followed by immediate detection/analysis. Subsequent analysis could include proteasomal degradation, ubiquitin modification site mapping (by mass spectrometry), and the effect of ubiquitin modification on enzyme interactions, activity and function, Ubiquitinylation of proteins of interest from cell or tissue extracts, Modification of proteins using ubiquitin derivatives or ubiquitin mutants for improved detection, analysis or investigation of alternative (non-proteasomal) ubiquitin signaling pathways.
Ordering information: Quantity: Provides sufficient material for 20 x 50μl reactions. Contents: HeLa S100 fraction (Prod. No. BML-SW8750)1 mg (200 µl); 5mg/ml10x Ubiquitin solution (UB) (Prod. No. BML-KW8795)100 µl; 1mg/ml10x ATP-(Energy) regeneration solutions (ATP-ERS) (Prod. No. BML-EW9810)100 µl10x Ubiquitinylation buffer (Prod. No. BML-KW9885a)100 µlUbiquitin aldehyde (inhibitor) (Prod. No. BML-UW8450)50 µl; 1mg/mlUbiquitin-conjugate specific antibody solution (FK2) (Prod. No. BML-KW8810)10 µl, Mono- and polyubiquitinylated proteins, multi-ubiquitin chains, mouse mAb (clone FK2)
Customers Who Bought This Also Bought