To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Tag sequence:HIS Tag
- Protein/peptide name:UbcH12
- Purity:>95% (SDS-PAGE)
- Formulation:Liquid. In HEPES, pH 7.5, containing 100mM NaCl, 1mM DTT, 10% glycerol.
- Cat. no.:89165-342
- Supplier No.:UW9145-0100
Spécifications
A propos de cet article
Produced in E. coli.
NEDD8 (neural precursor cell expressed developmentally downregulated gene 8) is an ubiquitin-like protein, with approx. 60% identity to ubiquitin. Conjugation of mature NEDD8 (via its exposed C-terminal glycine76 residue) to specific lysine residues on a limited number of cellular target proteins via isopeptide bonds, allows NEDD8 to play a critical regulatory role in cell proliferation and development, a process closely associated to the ubiquitin-proteasome pathway. The NEDD8 conjugation pathway proceeds by a mechanism analogous to that of the ubiquitin conjugation cascade, consisting of a dedicated NEDD8 E1 activating enzyme (APP-BP1/Uba3), E2 conjugating enzyme (UbcH12) and substrate specific E3 ligases. An increasing number of substrates for NEDDylation, including the tumor suppressors p53 and VHL, have been identified in addition to members of the well characterised cullin family of proteins, that play a structural role in ubiquitin E3 ligase complexes such as SCF. Cullin NEDDylation in SCF complexes is facilitated by the RING domain containing Roc1 SCF subunit, which is thought to act as a NEDD8 E3 ligase, and enhances the complex's ubiquitin E3 ligase activity. NEDDylation of p53 is mediated by its ubiquitin E3 ligase Mdm2, providing an additional control mechanism for inhibition of its transcriptional activity. Reconstitution of the NEDD8 conjugation pathway in vitro using recombinant proteins is readily achieved and has been used to investigate a number of NEDD8 modification systems.