ContactUsLinkComponent
SUMOylation Kit, Enzo Life Sciences
SUMOylation Kit, Enzo Life Sciences
# de catalogue 89165-294
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
SUMOylation Kit, Enzo Life Sciences
# de catalogue 89165-294
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    SUMOylation Kit
  • Size:
    1 Kit
  • Cat. no.:
    89165-294
  • No. of Reactions:
    20 assays
  • Supplier No.:
    UW8955-0001
Spécifications
A propos de cet article
Kit for generation of SUMOylated proteins in vitro.
  • SUMO-modification of specific proteins in vitro
  • Demonstrate novel proteins are potential targets for SUMOylation under in vitro conditions.
  • Generate substrates for deSUMOylating enzymes, such as SENP1 and SENP2
  • Facilitate or enhance SUMOylation of specific target proteins, particularly under conditions/enzyme concentrations that more closely represent those in vivo
  • SUMOylation of proteins in cell lysates or crude fractions/preparations to facilitate investigation of their role/function in complex solutions
  • Demonstrate SUMOylation of known proteins in specific lysates (confirm with target protein specific antibodies)
  • Use of cell lysate or crude fractions/preparations as source of SUMO E3 ligases to facilitate SUMOylation of purified target proteins in the presence of kit components
This kit provides a means of generating SUMOylated proteins in vitro, by covalent linkage of the carboxy-terminal of SUMO-1, -2 or -3 to specific lysine residues on the target protein via isopeptide bonds, using the SUMOylation enzyme cascade
Suggested uses: For SUMO-modification of specific proteins in vitro, To demonstrate that novel proteins are potential targets for SUMOylation under in vitro conditions, To generate substrates for deSUMOylating enzymes, such as SENP1 and SENP2, To test proteins for SUMO E3 ligase activity.
Note: Protocol provided for application 1. Assay set-up can be readily modified for alternative applications by inclusion, omission or substitution of specific components.
A short sequence containing the consensus ?-K-X-D/E (where lysine is the amino acid modified, ? is a large hydrophobic residue and X is any amino acid residue) is thought to be necessary for this in vitro protein SUMOylation to occur, however SUMOylation has also been observed in cases where the consensus site is not conserved. A control target protein is provided together with all other necessary components. SUMO specific antibodies are provided for detection of SUMOylated proteins via SDS-PAGE and Western blotting. Provides sufficient material for 20 x 20µL reactions.
Delivery: Kit contains, 20X SUMO Activating Enzyme Solution (SUMO E1) (20 µL), 20X SUMO Conjugating Enzyme Solution (SUMO E2) (20 µL), 20X SUMO Enzyme Solutions (SUMO1, SUMO2, SUMO3) (20 µL each), 10X SUMOylation Buffer (40 µL), 20X Control RanGAP1 SUMOylation Target Protein Solution (RG1) (20 µL), 20X Mg-ATP Solution (20 µL), SUMO1 Antibody Solution (25 µL; SUMO1 (CT) rabbit polyclonal antibody), SUMO2/3 Antibody Solution (25 µL; SUMO2/3 (NT) rabbit polyclonal antibody).
Customers Who Bought This Also Bought