ContactUsLinkComponent
Auto-Ubiquitinylation Kit, Enzo Life Sciences
Auto-Ubiquitinylation Kit, Enzo Life Sciences
# de catalogue 89165-248
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Auto-Ubiquitinylation Kit 1 Kit
Auto-Ubiquitinylation Kit, Enzo Life Sciences
# de catalogue 89165-248
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Auto-Ubiquitinylation Kit
  • Size:
    1 Kit
  • Storage Temperature:
    -80 °C
  • No. of Assays/Kit:
    10
  • Supplier No.:
    UW0970-0001
Spécifications
A propos de cet article
This E3 ligase auto-ubiquitinylation kit enables proteins to be tested for ubiquitin E3 ligase activity through assessment of their ability to under auto-ubiquitinylation
Utilizing the first three steps in the ubiquitin cascade the kit facilitates ubiquitinylation of known or putative E3 ligase enzymes followed by Western blot analysis using the highly sensitive reagents provided or using antibodies to the specific protein of interest (user supplied). A high integrity ubiquitin E3 ligase enzyme is also provided for use as a positive control.
Delivery: Kit contains 20X Ubiquitin Activating Enzyme Solution (E1) (25µL), 20X Ubiquitin Conjugating Enzyme Solution (E2) (25µL), 10X Ubiquitin Solution (Ub) (25µL), 20X Control Ubiquitin Ligase Solution (E3) (25µL), 20X Mg-ATP Solution (25µL), 10X Ub E3 ligase Buffer (50µL), Ubiquitin antibody solution (25µL).
Customers Who Bought This Also Bought