ContactUsLinkComponent
NEDDylation Kit, Enzo Life Sciences
NEDDylation Kit, Enzo Life Sciences
# de catalogue 89165-182
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
NEDDylation Kit, Enzo Life Sciences
# de catalogue 89165-182
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    NEDDylation Kit
  • Size:
    1 Kit
  • Storage temperature:
    -80 °C
  • Cat. no.:
    89165-182
  • No. of Reactions:
    20
  • Supplier No.:
    UW0590-0001
Spécifications
A propos de cet article
This kit provides the means of generating a thioester-linked NEDD8-conjugated E2 enzyme, utilizing the first two steps in the NEDD8 cascade, for use in the NEDDylation of E3 ligases and target substrate proteins (user supplied)
  • This kit provides sufficient material for 20 x 20µL reactions
  • Generate NEDD8-E2 thioesters for use in the in vitro NEDDylation of target proteins
  • Demonstrate novel proteins are potential targets for NEDDylation
  • Study NEDD8 activation (E1) and conjugation (E2) steps in the NEDDylation cascade
  • Generate substrates for deNEDDylating enzymes
  • Test proteins for NEDD8 E3 ligase activity
  • Avoid freeze/thaw cycles
Note: This Kit does not contain any NEDD8 E3 ligases.
A NEDD8-specific antibody is provided for detection of NEDDylated proteins via SDS-PAGE and Western blotting.
Delivery: NEDD8 Activating Enzyme Solution (E1), UbcH12 Solution (E2), NEDD8 Solution NEDDylation Buffer Mg-ATP Solution, NEDD8 Antibody Solution, Non-reducing Gel Loading Buffer.
Customers Who Bought This Also Bought