To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Size:1 mg
- Protein/peptide name:SUMO-1 (93-97)-AMCA
- Purity:>95% (HPLC)
- Formulation:Lyophilized solid.
- Cat. no.:89165-164
- Supplier No.:UW0500-1000
Spécifications
A propos de cet article
SUMO1-AMCA is a synthetic AMCA-peptide substrate based on the C-terminal residues of SUMO1 and is a useful fluorogenic substrate for deSUMOylating enzymes, including SENP1 (BML-UW9760). It has particular utility for the study of deSUMOylating activity where continuous monitoring of activity is essential. It provides an alternative substrate to SUMO1-AMC.