ContactUsLinkComponent
Nuclear extract from HeLa cells, Enzo Life Sciences
Nuclear extract from HeLa cells, Enzo Life Sciences
# de catalogue 89160-320
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Nuclear Extract From Hela Cells 100 ul
Nuclear extract from HeLa cells, Enzo Life Sciences
# de catalogue 89160-320
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    HeLa Nuclear Extract
  • Size:
    100 µl
  • Cat. no.:
    89160-320
  • Supplier No.:
    KI140-0100
Spécifications
A propos de cet article
Prepared by high salt extraction of HeLa nuclei (human cervical cancer cell line), this extract is a rich source of HDAC activity, useful for inhibitor screening or immunoprecipitation of HDAC containing complexes. Typically, 0.5 µl or less per well is enough for assay with the HDAC FLUOR DE LYS® fluorometric activity assay.
Formulation: Liquid in 0.1 M KCl, 20 mM HEPES/NaOH, pH 7.9, 20% (v/v) glycerol, 0.2 mM EDTA, 0.5 mM DTT, 0.5 mM PMSF.
Customers Who Bought This Also Bought