To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Size:50 µl
- Protein/peptide name:FLUOR DE LYS®-Deacetylase Substrate
- Purity:95% (HPLC)
- Formulation:Supplied as a 50 mM solution (50 µl) in dimethylsulfoxide (DMSO).
- Cat. no.:89160-282
- Supplier No.:KI104-0050
Spécifications
A propos de cet article
Fluor de Lys® Substrate is a highly sensitive and convenient alternative to radiolabeled, acetylated histones or peptide/HPLC methods for the assay of histone deacetylases. The Fluor de Lys® Substrate, which comprises an acetylated lysine side chain, is incubated with a sample containing HDAC activity (nuclear or cellular extract, purified enzyme, bead-bound immunocomplex, etc.). Deacetylation of the substrate sensitizes the substrate so that, in a second step, treatment with the Fluor de Lys® Developer produces a fluorophore. Histone deacetylases from all known classes have been successfully assayed with the Fluor de Lys® Substrate. These include human HDACs 1, 2, 3 and 8 (class I), human HDACs 4 and 9 (class II) and the sirtuins yeast Sir2, human SIRT1 and human SIRT2 (class III; reaction must include the sirtuin substrate NAD+). The substrate is cell-permeable and serves as the basis for the HDAC Cellular Activity Assay Kit (Prod. No. BML-AK503). NOTE: Must be used in conjunction with Fluor de Lys® Developer (Prod. No. BML-KI105). Sufficient for 100-1000 assays of HDACs or sirtuins (50 µl reaction volume, 50-500 µM (substrate).